siRNA Details
| Gene Symbol: Idh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173011.2 | NP_766599.2 | 5 | 695 | ||
| GenBank mRNA | AF212319.1 | - | 5 | 666 | ||
| AK088110.1 | - | 5 | 667 | |||
| AK145753.1 | - | 5 | 640 | |||
| AK146564.1 | - | 5 | 678 | |||
| AK161640.1 | - | 5 | 679 | |||
| AK169395.1 | - | 5 | 692 | |||
| AK205677.1 | - | 1 | 50 | |||
| BC060030.1 | - | 5 | 683 | 6515657 | ||

Idh2
Genomic Map