siRNA Details
| Gene Symbol: Ttll13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177765.3 | NP_808433.2 | 9 | 1430 | ||
| GenBank mRNA | AK029565.1 | - | 9 | 1429 | ||
| AK161519.1 | - | 9 | 1253 | |||
| AM690757.1 | - | 8 | 1131 | |||
| BC145749.1 | - | 8 | 1038 | |||

Ttll13
Genomic Map