siRNA Details
| Gene Symbol: Fam155a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173446.2 | NP_775622.1 | 2 | 1022 | ||
| GenBank mRNA | AK032202.1 | - | 2 | 1022 | ||
| AK075681.1 | - | 2 | 153 | |||
| AK162130.1 | - | 2 | 559 | |||
| BC117535.1 | - | 2 | 1020 | 40106181 | ||

Fam155a
Genomic Map