siRNA Details
| Gene Symbol: Vat1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173016.3 | NP_766604.2 | 8 | 1185 | ||
| GenBank mRNA | AK035009.1 | - | 1 | 1776 | ||
| AK122527.1 | - | 8 | 1185 | |||
| BC056927.1 | - | 8 | 1195 | 5719211 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vat1l| Gene Symbol: Vat1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173016.3 | NP_766604.2 | 8 | 1185 | ||
| GenBank mRNA | AK035009.1 | - | 1 | 1776 | ||
| AK122527.1 | - | 8 | 1185 | |||
| BC056927.1 | - | 8 | 1195 | 5719211 | ||