siRNA Details
| Gene Symbol: Nlrx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163742.1 | NP_001157214.1 | 5 | 747 | ||
| NM_001163743.1 | NP_001157215.1 | 5 | 848 | |||
| NM_178420.3 | NP_848507.2 | 5 | 836 | |||
| GenBank mRNA | AK089260.1 | - | 5 | 840 | ||
| AK141296.1 | - | 5 | 817 | |||
| AK145780.1 | - | 5 | 858 | |||
| AK145988.1 | - | 5 | 837 | |||
| AK166689.1 | - | 5 | 674 | |||
| AK170088.1 | - | 5 | 791 | |||
| BC034204.1 | - | 4 | 630 | |||
| BC050054.1 | - | 5 | 743 | 30012053 | ||

Nlrx1
Genomic Map