siRNA Details
| Gene Symbol: Nlrx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163742.1 | NP_001157214.1 | 3 | 250 | ||
| NM_001163743.1 | NP_001157215.1 | 3 | 351 | |||
| NM_178420.3 | NP_848507.2 | 3 | 339 | |||
| GenBank mRNA | AK089260.1 | - | 3 | 343 | ||
| AK141296.1 | - | 3 | 320 | |||
| AK145780.1 | - | 3 | 360 | |||
| AK145988.1 | - | 3 | 340 | |||
| AK166689.1 | - | 3 | 177 | |||
| AK170088.1 | - | 3 | 294 | |||
| BC050054.1 | - | 3 | 246 | 30012053 | ||
| BY196718.1 | - | 3 | 250 | |||

Nlrx1
Genomic Map