siRNA Details
| Gene Symbol: Clpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044389.2 | NP_001037854.1 | 6 | 959 | ||
| NM_011802.3 | NP_035932.2 | 7 | 1001 | |||
| GenBank mRNA | AF134983.1 | - | 7 | 952 | ||
| AJ276991.1 | - | 7 | 883 | |||
| AK087848.1 | - | 7 | 955 | |||
| BC061153.1 | - | 6 | 917 | 6593118 | ||

Clpx
Genomic Map