siRNA Details
Gene Symbol: Pfkfb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173019.5 | NP_766607.3 | 13, 14 | 1463 | ||
GenBank mRNA | AK048999.1 | - | 5, 6 | 577 | ||
AK155172.1 | - | 13, 14 | 1401 | |||
AK213190.1 | - | 1, 2 | 36 | |||
AY646092.2 | - | 13, 14 | 1349 | |||
AY707862.1 | - | 13, 14 | 1366 | |||
AY756065.1 | - | 13, 14 | 1456 | |||
AY756066.1 | - | 13, 14 | 1456 | |||
AY756067.1 | - | 13, 14 | 1327 | |||
BC057594.1 | - | 13, 14 | 1415 | 6810667 | ||
DQ350638.1 | - | 14, 15 | 1435 | |||
EU571215.1 | - | 13, 14 | 1447 | |||
EU571216.1 | - | 15, 16 | 1622 | |||
EU571217.1 | - | 13, 14 | 1590 |
