siRNA Details
| Gene Symbol: G3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013716.2 | NP_038744.1 | 11 | 1240 | ||
| GenBank mRNA | AB001927.1 | - | 11 | 1240 | ||
| AK220230.1 | - | 11 | 1294 | |||
| BC021156.1 | - | 11 | 1252 | 4020362 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

G3bp1| Gene Symbol: G3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013716.2 | NP_038744.1 | 11 | 1240 | ||
| GenBank mRNA | AB001927.1 | - | 11 | 1240 | ||
| AK220230.1 | - | 11 | 1294 | |||
| BC021156.1 | - | 11 | 1252 | 4020362 | ||