siRNA Details
| Gene Symbol: Nit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242580.1 | NP_001229509.1 | 6 | 763 | ||
| NM_012049.2 | NP_036179.1 | 6 | 731 | |||
| GenBank mRNA | AF069988.1 | - | 6 | 712 | ||
| AK002269.1 | - | 6 | 771 | |||
| AK004988.2 | - | 724 | ||||
| AK032189.1 | - | 1414 | ||||
| AK213449.1 | - | 1 | 49 | |||

Nit1
Genomic Map