siRNA Details
| Gene Symbol: Aoah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012054.4 | NP_036184.1 | 16 | 1559 | ||
| GenBank mRNA | AK084452.1 | - | 16 | 1554 | ||
| AK085343.1 | - | 16 | 1531 | |||
| AK133301.1 | - | 16 | 1500 | |||
| AK170293.1 | - | 15 | 1095 | |||
| AK171572.1 | - | 16 | 1378 | |||
| BC150767.1 | - | 16 | 1353 | 9087678 | ||
| CT010242.1 | - | 16 | 1278 | |||

Aoah
Genomic Map