siRNA Details
| Gene Symbol: Tcirg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136091.2 | NP_001129563.1 | 15 | 1851 | ||
| NM_001167784.1 | NP_001161256.1 | 15 | 2196 | |||
| NM_016921.3 | NP_058617.3 | 15 | 1892 | |||
| GenBank mRNA | AB022322.1 | - | 15 | 1758 | ||
| AF218253.1 | - | 15 | 1830 | |||
| AF218254.1 | - | 8 | 1513 | |||
| AK004094.1 | - | 6 | 702 | |||
| AK155140.1 | - | 15 | 2199 | |||
| AK183360.1 | - | 1 | 37 | |||
| BC006761.1 | - | 15 | 1892 | 3257437 | ||
| BC085234.1 | - | 15 | 1752 | 5322429 | ||

Tcirg1
Genomic Map