siRNA Details
Gene Symbol: Taf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290729.1 | NP_001277658.1 | 7 | 1264 | ||
GenBank mRNA | AB299229.1 | - | 7 | 1191 | ||
AK143571.1 | - | 7 | 1241 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Taf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290729.1 | NP_001277658.1 | 7 | 1264 | ||
GenBank mRNA | AB299229.1 | - | 7 | 1191 | ||
AK143571.1 | - | 7 | 1241 |