siRNA Details
| Gene Symbol: Mthfd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170785.1 | NP_001164256.1 | 11 | 1324 | ||
| NM_001170786.1 | NP_001164257.1 | 11 | 1324 | |||
| NM_172308.4 | NP_758512.3 | 11 | 1324 | |||
| GenBank mRNA | AK028211.1 | - | 11 | 1275 | ||
| AK036284.1 | - | 11 | 1270 | |||
| AK038579.1 | - | 11 | 1282 | |||
| AK075931.1 | - | 11 | 1250 | |||
| AK160025.1 | - | 11 | 1256 | |||
| AK197409.1 | - | 2 | 149 | |||
| BC030437.1 | - | 7 | 658 | 5374413 | ||
| BC049936.1 | - | 7 | 647 | |||

Mthfd1l
Genomic Map