siRNA Details
| Gene Symbol: Klhdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178253.5 | NP_839984.1 | 6 | 638 | ||
| GenBank mRNA | AY238938.1 | - | 6 | 638 | ||
| BC119567.2 | - | 6 | 616 | 40057793 | ||
| BC119568.2 | - | 6 | 622 | 40057794 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhdc1| Gene Symbol: Klhdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178253.5 | NP_839984.1 | 6 | 638 | ||
| GenBank mRNA | AY238938.1 | - | 6 | 638 | ||
| BC119567.2 | - | 6 | 616 | 40057793 | ||
| BC119568.2 | - | 6 | 622 | 40057794 | ||