siRNA Details
| Gene Symbol: Eif2ak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177806.1 | NP_001171277.1 | 27 | 3648 | ||
| NM_013719.3 | NP_038747.2 | 28 | 3952 | |||
| GenBank mRNA | AJ243533.1 | - | 28 | 3952 | ||
| AK011380.1 | - | 1 | 38 | |||
| AK129334.1 | - | 23 | 3302 | |||
| BC072637.1 | - | 27 | 3648 | 30354433 | ||
| BF453236.1 | - | 6 | 566 | |||
| BF462783.1 | - | 1 | 123 | |||

Eif2ak4
Genomic Map