siRNA Details
| Gene Symbol: Adamts16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172053.2 | NP_742050.2 | 5 | 1101 | ||
| GenBank mRNA | AK031314.1 | - | 5 | 1067 | ||
| AK173338.1 | - | 5 | 1037 | |||
| BC034739.1 | - | 5 | 1114 | 4951547 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adamts16| Gene Symbol: Adamts16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172053.2 | NP_742050.2 | 5 | 1101 | ||
| GenBank mRNA | AK031314.1 | - | 5 | 1067 | ||
| AK173338.1 | - | 5 | 1037 | |||
| BC034739.1 | - | 5 | 1114 | 4951547 | ||