siRNA Details
| Gene Symbol: Ankdd1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042714.1 | NP_001036179.1 | 9 | 943 | ||
| GenBank mRNA | AK033386.1 | - | 3 | 183 | ||
| AK033960.1 | - | 2 | 137 | |||
| AK084814.1 | - | 2 | 104 | |||
| BC112417.1 | - | 9 | 943 | 40061167 | ||

Ankdd1b
Genomic Map