siRNA Details
| Gene Symbol: Rp1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146246.3 | NP_666358.2 | 4 | 5317 | ||
| GenBank mRNA | AJ491325.1 | - | 3 | 5119 | ||
| AY168340.1 | - | 3 | 5098 | |||
| BC031365.1 | - | 1 | 1455 | |||
| BC049924.1 | - | 1 | 1455 | |||
| BC129887.1 | - | 4 | 5242 | 40111348 | ||

Rp1l1
Genomic Map