siRNA Details
| Gene Symbol: BC024139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142968.1 | NP_001136440.1 | 12 | 1792 | ||
| GenBank mRNA | AK078032.1 | - | 12 | 1780 | ||
| AK081549.1 | - | 12 | 1794 | |||
| BC060242.1 | - | 12 | 1832 | |||
| BC063071.1 | - | 7 | 1227 | 5694464 | ||

BC024139
Genomic Map