siRNA Details
| Gene Symbol: Ip6k3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173027.2 | NP_766615.1 | 6 | 1364 | ||
| GenBank mRNA | AK052857.1 | - | 6 | 1365 | ||
| BC141263.1 | - | 6 | 1258 | 9055875 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ip6k3| Gene Symbol: Ip6k3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173027.2 | NP_766615.1 | 6 | 1364 | ||
| GenBank mRNA | AK052857.1 | - | 6 | 1365 | ||
| BC141263.1 | - | 6 | 1258 | 9055875 | ||