siRNA Details
| Gene Symbol: Vps13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173028.4 | NP_766616.2 | 52 | 7381 | ||
| GenBank mRNA | AB115421.1 | - | 52 | 7376 | ||
| AK029649.1 | - | 6 | 1136 | |||
| AK030073.1 | - | 5 | 605 | |||
| AK135983.1 | - | 9 | 1535 | |||
| AK220362.1 | - | 5 | 798 | |||
| BC062130.1 | - | 8 | 1279 | |||
| BC096768.1 | - | 6 | 977 | |||
| BC145012.1 | - | 54 | 7651 | |||

Vps13a
Genomic Map