siRNA Details
Gene Symbol: Agbl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178755.3 | NP_848870.2 | 9 | 1603 | ||
GenBank mRNA | AK029744.1 | - | 9 | 1636 | ||
AK029932.1 | - | 9 | 1604 | |||
AK030138.1 | - | 9 | 1591 | |||
AK030182.1 | - | 9 | 1650 | |||
AK040259.1 | - | 9 | 1539 | |||
AK041286.1 | - | 1 | 12 | |||
AK079691.1 | - | 9 | 1540 | |||
DQ867026.1 | - | 9 | 1636 | |||
DQ867027.1 | - | 9 | 1636 | |||
DQ867028.1 | - | 9 | 1636 | |||
DQ867029.1 | - | 9 | 1636 | |||
DQ867030.1 | - | 8 | 1582 |
