siRNA Details
| Gene Symbol: Tbck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034533.1 | - | 10 | 1060 | ||
| AK052841.1 | - | 10 | 1072 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbck| Gene Symbol: Tbck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034533.1 | - | 10 | 1060 | ||
| AK052841.1 | - | 10 | 1072 | |||