siRNA Details
| Gene Symbol: Tbck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034533.1 | - | 20 | 2077 | ||
| CN720994.1 | - | 2 | 139 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbck| Gene Symbol: Tbck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034533.1 | - | 20 | 2077 | ||
| CN720994.1 | - | 2 | 139 | |||