siRNA Details
| Gene Symbol: Tstd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173033.3 | NP_766621.3 | 8 | 1135 | ||
| GenBank mRNA | AK028258.1 | - | 8 | 1078 | ||
| AK155456.1 | - | 8 | 1082 | |||
| BC057893.1 | - | 8 | 1070 | 4978941 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tstd2| Gene Symbol: Tstd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173033.3 | NP_766621.3 | 8 | 1135 | ||
| GenBank mRNA | AK028258.1 | - | 8 | 1078 | ||
| AK155456.1 | - | 8 | 1082 | |||
| BC057893.1 | - | 8 | 1070 | 4978941 | ||