siRNA Details
Gene Symbol: Lppr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178756.4 | NP_848871.1 | 7, 8 | 1135 | ||
GenBank mRNA | AK034251.1 | - | 7, 8 | 1230 | ||
AK043762.1 | - | 7, 8 | 1037 | |||
AK047614.1 | - | 7, 8 | 1065 | |||
AK087439.1 | - | 7, 8 | 1286 | |||
AK087514.1 | - | 7, 8 | 1136 | |||
AK160897.1 | - | 7, 8 | 1262 | |||
AY345342.1 | - | 7, 8 | 1135 | |||
BC096762.1 | - | 7, 8 | 1111 | 30094664 |
