siRNA Details
| Gene Symbol: Nrk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013724.2 | NP_038752.2 | 1, 2 | 380 | ||
| GenBank mRNA | AB020741.1 | - | 1, 2 | 305 | ||
| AB035267.1 | - | 1, 2 | 58 | |||
| AK041377.1 | - | 1, 2 | 380 | |||
| BC068311.1 | - | 1, 2 | 355 | |||
| BC139029.1 | - | 1, 2 | 147 | 8862051 | ||
| BC139031.1 | - | 1, 2 | 147 | 8862053 | ||

Nrk
Genomic Map