siRNA Details
| Gene Symbol: Dbf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190717.1 | NP_001177646.1 | 12 | 1943 | ||
| NM_013726.3 | NP_038754.1 | 12 | 1946 | |||
| GenBank mRNA | AJ003132.1 | - | 12 | 1836 | ||
| AK132403.1 | - | 12 | 1908 | |||
| AK161782.1 | - | 1 | 927 | |||
| BC138709.1 | - | 12 | 1764 | 8861731 | ||
| BC138710.1 | - | 12 | 1764 | 8861732 | ||
| BC145392.1 | - | 12 | 1761 | 9053950 | ||

Dbf4
Genomic Map