siRNA Details
| Gene Symbol: Zfp398 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027477.3 | NP_081753.1 | 6 | 1733 | ||
| NM_173034.3 | NP_766622.2 | 5 | 1337 | |||
| GenBank mRNA | AK078250.1 | - | 5 | 1304 | ||
| AK081169.1 | - | 6 | 1698 | |||
| AK089190.1 | - | 6 | 1702 | |||
| AK155461.1 | - | 6 | 1700 | |||
| BC112382.1 | - | 6 | 1659 | 40059618 | ||

Zfp398
Genomic Map