siRNA Details
| Gene Symbol: B3gnt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081167.1 | NP_001074636.1 | 2 | 185 | ||
| GenBank mRNA | BC039789.1 | - | 2 | 201 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B3gnt6| Gene Symbol: B3gnt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081167.1 | NP_001074636.1 | 2 | 185 | ||
| GenBank mRNA | BC039789.1 | - | 2 | 201 | ||