siRNA Details
| Gene Symbol: Plek2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013738.3 | NP_038766.1 | 9 | 960 | ||
| GenBank mRNA | AF157600.1 | - | 9 | 1006 | ||
| AF170564.1 | - | 9 | 972 | |||
| BC028902.1 | - | 9 | 960 | 4208341 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plek2| Gene Symbol: Plek2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013738.3 | NP_038766.1 | 9 | 960 | ||
| GenBank mRNA | AF157600.1 | - | 9 | 1006 | ||
| AF170564.1 | - | 9 | 972 | |||
| BC028902.1 | - | 9 | 960 | 4208341 | ||