siRNA Details
| Gene Symbol: Clnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013748.3 | NP_038776.3 | 11 | 694 | ||
| GenBank mRNA | AB021220.1 | - | 11 | 814 | ||
| AF187819.1 | - | 11 | 694 | |||
| AK082826.1 | - | 7 | 432 | |||
| BC125637.1 | - | 11 | 691 | 40130152 | ||
| BC125639.1 | - | 11 | 691 | 40130154 | ||

Clnk
Genomic Map