siRNA Details
| Gene Symbol: Ptdss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013782.4 | NP_038810.2 | 6 | 651 | ||
| GenBank mRNA | AF099053.1 | - | 6 | 535 | ||
| AK021249.1 | - | 6 | 616 | |||
| AK031906.1 | - | 6 | 637 | |||
| AK032686.1 | - | 6 | 616 | |||
| AK036368.1 | - | 6 | 652 | |||
| AK048026.1 | - | 6 | 634 | |||
| AK081275.1 | - | 6 | 618 | |||
| AK137415.1 | - | 611 | ||||
| BC009013.1 | - | 4 | 318 | |||
| BC053517.1 | - | 6 | 588 | 6315175 | ||

Ptdss2
Genomic Map