siRNA Details
| Gene Symbol: Dusp13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007268.1 | NP_001007269.1 | 7 | 1195 | ||
| NM_013849.3 | NP_038877.2 | 3 | 520 | |||
| GenBank mRNA | AB027003.1 | - | 2 | 301 | ||
| AF237620.1 | - | 3 | 503 | |||
| AK006247.1 | - | 3 | 521 | |||
| AY674052.1 | - | 7 | 1195 | |||
| BC100421.1 | - | 2 | 426 | 30919819 | ||

Dusp13
Genomic Map