siRNA Details
| Gene Symbol: Mmel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013783.2 | NP_038811.2 | 3 | 478 | ||
| GenBank mRNA | AF157105.1 | - | 3 | 424 | ||
| AF176569.1 | - | 3 | 475 | |||
| AF302076.1 | - | 2 | 168 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmel1| Gene Symbol: Mmel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013783.2 | NP_038811.2 | 3 | 478 | ||
| GenBank mRNA | AF157105.1 | - | 3 | 424 | ||
| AF176569.1 | - | 3 | 475 | |||
| AF302076.1 | - | 2 | 168 | |||