siRNA Details
| Gene Symbol: Mmel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013783.2 | NP_038811.2 | 11 | 1310 | ||
| GenBank mRNA | AF157105.1 | - | 11 | 1256 | ||
| AF157106.1 | - | 9 | 965 | |||
| AF176569.1 | - | 11 | 1307 | |||
| AF302075.1 | - | 9 | 931 | |||
| AF302076.1 | - | 10 | 1000 | |||
| AF302077.1 | - | 9 | 931 | |||
| AK154366.1 | - | 8 | 799 | |||
| BC137649.1 | - | 9 | 1083 | 8860668 | ||
| BC144804.1 | - | 9 | 1083 | 9053352 | ||

Mmel1
Genomic Map