siRNA Details
| Gene Symbol: Abca3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039581.2 | NP_001034670.1 | 23 | 3988 | ||
| NM_013855.3 | NP_038883.2 | 24 | 4061 | |||
| GenBank mRNA | AK089794.1 | - | 24 | 4005 | ||
| AK154698.1 | - | 24 | 3969 | |||
| AK171239.1 | - | 6 | 1102 | |||
| AY083616.1 | - | 24 | 3956 | |||
| BC042663.1 | - | 18 | 3003 | |||
| BC079617.1 | - | 18 | 2787 | 30362309 | ||
| CV784401.1 | - | 4 | 540 | |||

Abca3
Genomic Map