siRNA Details
| Gene Symbol: Abcb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021022.3 | NP_066302.2 | 23 | 2991 | ||
| GenBank mRNA | AF133903.1 | - | 23 | 2962 | ||
| AI116345.1 | - | 3 | 358 | |||
| AK144553.1 | - | 24 | 3066 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcb11| Gene Symbol: Abcb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021022.3 | NP_066302.2 | 23 | 2991 | ||
| GenBank mRNA | AF133903.1 | - | 23 | 2962 | ||
| AI116345.1 | - | 3 | 358 | |||
| AK144553.1 | - | 24 | 3066 | |||