siRNA Details
| Gene Symbol: Abcb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021022.3 | NP_066302.2 | 9 | 953 | ||
| GenBank mRNA | AF133903.1 | - | 9 | 924 | ||
| AK144553.1 | - | 10 | 1028 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcb11| Gene Symbol: Abcb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021022.3 | NP_066302.2 | 9 | 953 | ||
| GenBank mRNA | AF133903.1 | - | 9 | 924 | ||
| AK144553.1 | - | 10 | 1028 | |||