siRNA Details
Gene Symbol: Mkln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013791.2 | NP_038819.1 | 18 | 2193 | ||
GenBank mRNA | AK077541.1 | - | 18 | 2188 | ||
BC013703.1 | - | 18 | 2177 | 3668925 | ||
CF161946.1 | - | 6 | 533 | |||
U72194.1 | - | 18 | 2176 |
