siRNA Details
| Gene Symbol: Abcc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018795.2 | NP_061265.2 | 3 | 336 | ||
| GenBank mRNA | AB028737.1 | - | 3 | 342 | ||
| BC040400.1 | - | 3 | 344 | |||
| EF109740.1 | - | 3 | 336 | |||
| EF109741.1 | - | 3 | 336 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcc6| Gene Symbol: Abcc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018795.2 | NP_061265.2 | 3 | 336 | ||
| GenBank mRNA | AB028737.1 | - | 3 | 342 | ||
| BC040400.1 | - | 3 | 344 | |||
| EF109740.1 | - | 3 | 336 | |||
| EF109741.1 | - | 3 | 336 | |||