siRNA Details
| Gene Symbol: Trpa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177781.4 | NP_808449.1 | 8 | 1004 | ||
| GenBank mRNA | AK045771.1 | - | 8 | 1005 | ||
| AY231177.1 | - | 8 | 977 | |||
| BC120563.1 | - | 8 | 995 | 40129486 | ||
| BC131963.1 | - | 8 | 995 | 40130240 | ||
| KF460461.1 | - | 8 | 977 | |||

Trpa1
Genomic Map