siRNA Details
| Gene Symbol: Prex1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177782.3 | NP_808450.2 | 6 | 794 | ||
| GenBank mRNA | AK173168.1 | - | 6 | 794 | ||
| BC067047.1 | - | 4 | 697 | |||
| BC151074.1 | - | 6 | 742 | 9087987 | ||
| BC157940.1 | - | 6 | 742 | 9088120 | ||

Prex1
Genomic Map