siRNA Details
| Gene Symbol: Slc39a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012305.2 | NP_001012305.1 | 8 | 1533 | ||
| GenBank mRNA | AK134622.1 | - | 8 | 1534 | ||
| BC089362.1 | - | 8 | 1527 | 30628112 | ||
| BC113764.1 | - | 8 | 1516 | 40059402 | ||
| BC113765.1 | - | 8 | 1516 | 40059404 | ||

Slc39a12
Genomic Map