siRNA Details
Gene Symbol: Zfp385c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177790.4 | NP_808458.2 | 5 | 875 | ||
GenBank mRNA | AK044299.1 | - | 5 | 876 | ||
BC116726.1 | - | 5 | 836 | 40106534 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp385c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177790.4 | NP_808458.2 | 5 | 875 | ||
GenBank mRNA | AK044299.1 | - | 5 | 876 | ||
BC116726.1 | - | 5 | 836 | 40106534 |