siRNA Details
| Gene Symbol: Wfikkn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181819.2 | NP_861540.2 | 1 | 49 | ||
| GenBank mRNA | AY100450.1 | - | 1 | 191 | ||
| AY308804.1 | - | 1 | 19 | |||
| BC113144.1 | - | 1 | 19 | 40047070 | ||
| JQ080910.1 | - | 1 | 19 | |||

Wfikkn2
Genomic Map