siRNA Details
| Gene Symbol: Tango2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138583.2 | NP_613049.2 | 9 | 802 | ||
| GenBank mRNA | AK089239.1 | - | 9 | 784 | ||
| AK133588.1 | - | 9 | 785 | |||
| AK168595.1 | - | 8 | 781 | |||
| BC005445.1 | - | 9 | 882 | 3496749 | ||
| X74504.1 | - | 8 | 775 | |||

Tango2
Genomic Map