siRNA Details
| Gene Symbol: Adamts13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001322.2 | NP_001001322.1 | 21, 22 | 2877 | ||
| NM_001290463.1 | NP_001277392.1 | 21, 22 | 2793 | |||
| NM_001290464.1 | NP_001277393.1 | 21, 22 | 2877 | |||
| NM_001290465.1 | NP_001277394.1 | 20, 21 | 2850 | |||
| GenBank mRNA | AB071302.2 | - | 21, 22 | 2877 | ||
| AB112362.1 | - | 21, 22 | 2756 | |||
| AK157459.1 | - | 21, 22 | 2719 | |||
| EU034706.1 | - | 20, 21 | 2729 | |||

Adamts13
Genomic Map