siRNA Details
| Gene Symbol: Pcdh19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105245.1 | NP_001098715.1 | 1 | 3471 | ||
| NM_001105246.1 | NP_001098716.1 | 1 | 3471 | |||
| GenBank mRNA | AK046726.1 | - | 1 | 3321 | ||
| AK122492.1 | - | 1 | 1214 | |||
| BC118529.1 | - | 2 | 1660 | 40098492 | ||

Pcdh19
Genomic Map